Please login to your online account to display your discounted pricing

Thermo Scientific™ ABgene™ EasyStrip PCR Tubes


Each tube in the strip has an individually sealable cap to reduce the chance of sample contamination. PCR and QPCR setup and analysis are made much simpler with the unique design of these new tubes.

Manufacturer: thermo scientific  AB1502*

 View more versions of this product

Catalog No. AB1502

  • / Pack of 250 Each
 

Description & Specifications

Specifications

Description For PCR or QPCR
Color Clear
Shape Flat
Cap Type Ultra-Clear Flat Caps
Quantity 250 Strips
For Use With (Equipment) 0.2mL Cycler Blocks