missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Vinylcarbazole, 98+%

Manufacturer:  Alfa Aesar™ H5617430

 View more versions of this product

Catalog No. AAH5617430



Description

Description

Applications
9-Vinylcarbazole is used as a monomer in the production of poly(vinylcarbazole), a conductive polymer, in which conductivity is photon-dependent. It is also used in the photoreceptors of photocopiers.

Solubility
Soluble in acetonitrile.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

9-Vinylcarbazole
60°C to 65°C
1484-13-5
MFCD00004966
132988
Soluble in acetonitrile.
C=CN1C2=CC=CC=C2C3=CC=CC=C31
193.249
193.24
154°C to 155°C
250g
C14H11N
UN2811
1-vinylcarbazole, 9-vinyl-9h-carbazole, 9-vinylcarbazole, 9h-carbazole, 9-ethenyl, carbazole, 9-vinyl, n-vinyl carbazole, n-vinylcarbazole, poly 9-vinylcarbazole, poly-n-vinylcarbazole, vinylcarbazole
KKFHAJHLJHVUDM-UHFFFAOYSA-N
9-ethenylcarbazole
15143
≥98%

Chemical Identifiers

1484-13-5
193.249
KKFHAJHLJHVUDM-UHFFFAOYSA-N
15143
C=CN1C2=CC=CC=C2C3=CC=CC=C31
C14H11N
MFCD00004966
1-vinylcarbazole, 9-vinyl-9h-carbazole, 9-vinylcarbazole, 9h-carbazole, 9-ethenyl, carbazole, 9-vinyl, n-vinyl carbazole, n-vinylcarbazole, poly 9-vinylcarbazole, poly-n-vinylcarbazole, vinylcarbazole
9-ethenylcarbazole
Safety and Handling

Safety and Handling

GHS H Statement
H341-H302-H312-H315-H317
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P272-P280-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c

H300-H312-H315-H317-H319-H341

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 216-055-0

RTECSNumber : FE6350000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents