Please login to your online account to display your discounted pricing

9-(Trifluoroacetyl)anthracene, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 9-(Trifluoroacetyl)anthracene
Name Note 98%
CAS 53531-31-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C16H9F3O
Formula Weight 274.24
MDL Number MFCD00001258
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC203470010

  SDS
thermo fisher scientific
203470010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC203470050

  SDS
thermo fisher scientific
203470050
Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 274.24
Physical Form Powder
Quantity 1g
Color Yellow
EINECS Number 258-609-4
Melting Point 83° to 86°C
CAS Max % 100.0
CAS Min % 97.5
Packaging Glass bottle
Synonym 9-Anthryl trifluoromethyl ketone
Chemical Name or Material 9-(Trifluoroacetyl)anthracene
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C16H9F3O
CAS 53531-31-0
MDL Number MFCD00001258