Please login to your online account to display your discounted pricing

9-(Trifluoroacetyl)anthracene, 98%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  203470050

 View more versions of this product

Catalog No. AC203470050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 274.24
Quantity 5g
Color Yellow
EINECS Number 258-609-4
Melting Point 83° to 86°C
CAS Max % 100.0
CAS Min % 97.5
Chemical Name or Material 9-(Trifluoroacetyl)anthracene
Infrared Spectrum Authentic
Packaging Glass Bottle
Assay Percent Range 98%
MDL Number MFCD00001258
Synonym 9-Anthryl trifluoromethyl ketone
Name Note 98%
Molecular Formula C16H9F3O
CAS 53531-31-0

Specifications

Chemical Name or Material 9-(Trifluoroacetyl)anthracene
Name Note 98%
CAS 53531-31-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C16H9F3O
Formula Weight 274.24
MDL Number MFCD00001258
Synonym 9-Anthryl trifluoromethyl ketone