missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Phenylcarbazole-3-boronic acid, 98%

Manufacturer:  Alfa Aesar™ H64059MD

 View more versions of this product

Catalog No. AAH64059MD



Description

Description

Applications
9-Phenylcarbazole-3-boronic acid is used in the synthesis of alkyl-functionalized organic dyes.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

250mg
C18H14BNO2
10566519
Slightly soluble in water.
B(C1=CC2=C(C=C1)N(C3=CC=CC=C32)C4=CC=CC=C4)(O)O
287.125
287.13
98%
854952-58-2
MFCD12196936
3-borono-9-phenyl-9h-carbazole, 9-phenyl carbazole-3-boronic acid, 9-phenyl-3-carbazoleboronic acid, 9-phenyl-9h-carbazol-3-yl boronic acid, 9-phenyl-9h-carbazol-3-ylboronic acid, 9-phenylcarbazole-3-boronic acid, b-9-phenyl-9h-carbazol-3-yl boronic acid, g00061-watson-int, n-phenylcarbazole-3-boronic acid, pubchem23232
JWJQEUDGBZMPAX-UHFFFAOYSA-N
(9-phenylcarbazol-3-yl)boronic acid
51358450
Solid

Chemical Identifiers

854952-58-2
287.125
JWJQEUDGBZMPAX-UHFFFAOYSA-N
51358450
B(C1=CC2=C(C=C1)N(C3=CC=CC=C32)C4=CC=CC=C4)(O)O
C18H14BNO2
MFCD12196936
3-borono-9-phenyl-9h-carbazole, 9-phenyl carbazole-3-boronic acid, 9-phenyl-3-carbazoleboronic acid, 9-phenyl-9h-carbazol-3-yl boronic acid, 9-phenyl-9h-carbazol-3-ylboronic acid, 9-phenylcarbazole-3-boronic acid, b-9-phenyl-9h-carbazol-3-yl boronic acid, g00061-watson-int, n-phenylcarbazole-3-boronic acid, pubchem23232
(9-phenylcarbazol-3-yl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

Documents