Learn More
9-Phenylcarbazole-3-boronic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6405903
Description
9-Phenylcarbazole-3-boronic acid is used in the synthesis of alkyl-functionalized organic dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications9-Phenylcarbazole-3-boronic acid is used in the synthesis of alkyl-functionalized organic dyes.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications
9-Phenylcarbazole-3-boronic acid | |
C18H14BNO2 | |
1 g | |
9-phenyl-9h-carbazol-3-yl boronic acid, 9-phenyl-9h-carbazol-3-ylboronic acid, 9-phenyl carbazole-3-boronic acid, 9-phenyl-3-carbazoleboronic acid, 9-phenylcarbazole-3-boronic acid, b-9-phenyl-9h-carbazol-3-yl boronic acid, n-phenylcarbazole-3-boronic acid, 3-borono-9-phenyl-9h-carbazole, pubchem23232, g00061-watson-int | |
JWJQEUDGBZMPAX-UHFFFAOYSA-N | |
(9-phenylcarbazol-3-yl)boronic acid | |
51358450 | |
98% |
854952-58-2 | |
MFCD12196936 | |
10566519 | |
Slightly soluble in water. | |
OB(O)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1 | |
287.13 | |
287.13 | |
Solid |
Chemical Identifiers
854952-58-2 | |
287.13 | |
JWJQEUDGBZMPAX-UHFFFAOYSA-N | |
51358450 | |
OB(O)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1 |
C18H14BNO2 | |
MFCD12196936 | |
9-phenyl-9h-carbazol-3-yl boronic acid, 9-phenyl-9h-carbazol-3-ylboronic acid, 9-phenyl carbazole-3-boronic acid, 9-phenyl-3-carbazoleboronic acid, 9-phenylcarbazole-3-boronic acid, b-9-phenyl-9h-carbazol-3-yl boronic acid, n-phenylcarbazole-3-boronic acid, 3-borono-9-phenyl-9h-carbazole, pubchem23232, g00061-watson-int | |
(9-phenylcarbazol-3-yl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only