Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Phenylanthracene, 98%

Chemical Identifiers

Chemical Name or Material 9-Phenylanthracene
CAS 602-55-1
Formula Weight 254.33
MDL Number MFCD00001252
UN Number UN3077
Beilstein 1910570
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6441903

  SDS
alfa aesar™
H6441903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6441906

  SDS
alfa aesar™
H6441906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 254.33
Melting Point 153° to 155°C (lit.)
Quantity 1g
Beilstein 1910570
Sensitivity Light Sensitive
Color Off-white to pale brown
CAS 602-55-1
Boiling Point 417°C (lit.)
DOT Information Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Chemical Name or Material 9-Phenylanthracene
UN Number UN3077
MDL Number MFCD00001252
Assay 98%
Physical Form Solid