missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Phenyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole, TCI America™

Manufacturer:  TCI America P2350200MG

 View more versions of this product

Catalog No. P2350200MG



Specifications

Specifications

9-Phenyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
240°C
618442-57-2
9-Phenyl-9H-carbazole-3,6-diboronic Acid Bis(pinacol) Ester
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(C4=C3C=C(C=C4)B5OC(C(O5)(C)C)(C)C)C6=CC=CC=C6
495.233
495.23
≥98.0% (HPLC,N)
White-Yellow
200mg
C30H35B2NO4
BMKVLWGCSCKZTD-UHFFFAOYSA-N
9-phenyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
12177223
Crystal-Powder at 20°C

Chemical Identifiers

618442-57-2
495.233
9-Phenyl-9H-carbazole-3,6-diboronic Acid Bis(pinacol) Ester
9-phenyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
C30H35B2NO4
BMKVLWGCSCKZTD-UHFFFAOYSA-N
12177223
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(C4=C3C=C(C=C4)B5OC(C(O5)(C)C)(C)C)C6=CC=CC=C6
Safety and Handling

Safety and Handling

TSCA : No

Documents