missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Phenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole, TCI America™

Manufacturer:  TCI America P2364200MG

 View more versions of this product

Catalog No. P2364200MG



Specifications

Specifications

9-Phenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
149°C
1246669-45-3
C24H24BNO2
9-Phenylcarbazole-2-boronic Acid Pinacol Ester, 4,4,5,5-Tetramethyl-2-(9-phenylcarbazol-2-yl)-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4N3C5=CC=CC=C5
369.271
369.27
White
200mg
≥98.0% (GC)
MFCD28129441
WUXIRZBTTICLCG-UHFFFAOYSA-N
9-phenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
59858738
Crystal-Powder at 20°C

Chemical Identifiers

1246669-45-3
369.271
WUXIRZBTTICLCG-UHFFFAOYSA-N
59858738
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4N3C5=CC=CC=C5
C24H24BNO2
MFCD28129441
9-Phenylcarbazole-2-boronic Acid Pinacol Ester, 4,4,5,5-Tetramethyl-2-(9-phenylcarbazol-2-yl)-1,3,2-dioxaborolane
9-phenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Safety and Handling

Safety and Handling

TSCA : No

Documents