Please login to your online account to display your discounted pricing

9-Phenanthrol, 60%, Tech., ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  198950050

 View more versions of this product

Catalog No. AC198950050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Color Brown
Quantity 5g
Chemical Name or Material 9-Phenanthrol
CAS 484-17-3
CAS Min % 55.0
CAS Max % 100.0
Purity Grade Technical
Physical Form Brown powder
Assay 55% min.
Assay Percent Range 60%
Molecular Formula C14H10O
Formula Weight 194.23g/mol
MDL Number MFCD00082298
Beilstein 06, 706
Synonym 9-Hydroxyphenanthrene

Specifications

Chemical Name or Material 9-Phenanthrol
CAS 484-17-3
CAS Min % 55.0
CAS Max % 100.0
Purity Grade Technical
Assay 55% min.
Assay Percent Range 60%
Molecular Formula C14H10O
Formula Weight 194.23g/mol
MDL Number MFCD00082298
Synonym 9-Hydroxyphenanthrene