Please login to your online account to display your discounted pricing

9-Phenanthrol, 60%, Tech., ACROS Organics™

Chemical Identifiers

Chemical Name or Material 9-Phenanthrol
CAS 484-17-3
CAS Min % 55.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥55%
Assay Percent Range 60%
Molecular Formula C14H10O
Formula Weight 194.23
MDL Number MFCD00082298
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC198950010

  SDS
fisher scientific acros
198950010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥55%
Purity Grade Technical Grade
Physical Form Powder
Formula Weight 194.23
Quantity 1g
Color Brown
Beilstein 06, 706
CAS Max % 100.0
CAS Min % 55.0
Molecular Formula C14H10O
CAS 484-17-3
Assay Percent Range 60%
Chemical Name or Material 9-Phenanthrol
MDL Number MFCD00082298
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym 9-Hydroxyphenanthrene

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.