missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-(p-Tolyl)carbazole, TCI America™

Manufacturer:  TCI America T22275G

 View more versions of this product

Catalog No. T22275G



Specifications

Specifications

9-(p-Tolyl)carbazole
107°C
19264-73-4
OZVXBXKVMMIGDV-UHFFFAOYSA-N
9-(4-methylphenyl)carbazole
15798651
Crystal-Powder at 20°C
Yellow
5g
C19H15N
CC1=CC=C(C=C1)N2C3=CC=CC=C3C4=CC=CC=C42
257.336
257.34
≥98.0% (GC)

Chemical Identifiers

19264-73-4
257.336
15798651
CC1=CC=C(C=C1)N2C3=CC=CC=C3C4=CC=CC=C42
C19H15N
OZVXBXKVMMIGDV-UHFFFAOYSA-N
9-(4-methylphenyl)carbazole
Safety and Handling

Safety and Handling

TSCA : No

Documents