missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Octadecynoic acid, 98%

$73.26 - $225.15

Chemical Identifiers

CAS 506-24-1
Molecular Formula C18H32O2
Molecular Weight (g/mol) 280.452
MDL Number MFCD00014386
InChI Key RGTIBVZDHOMOKC-UHFFFAOYSA-N
Synonym stearolic acid, 9-octadecynoic acid, 9-stearolic acid, delta 9-octadecynoic acid, stearolsaeure, octadec-9-insaeure, octadeca-9-ynoic acid, 9-odya, d09qia, 9-oda
PubChem CID 68167
ChEBI CHEBI:28801
IUPAC Name octadec-9-ynoic acid
SMILES CCCCCCCCC#CCCCCCCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0129403 View Documents Alfa Aesar™
L0129403
1g Each for $73.26
 
AAL0129406 View Documents Alfa Aesar™
L0129406
5g Each for $225.15
 
Description

Description

Applications
9-Octadecynoic acid, is used as an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Chemical Identifiers

506-24-1
280.452
RGTIBVZDHOMOKC-UHFFFAOYSA-N
68167
octadec-9-ynoic acid
C18H32O2
MFCD00014386
stearolic acid, 9-octadecynoic acid, 9-stearolic acid, delta 9-octadecynoic acid, stearolsaeure, octadec-9-insaeure, octadeca-9-ynoic acid, 9-odya, d09qia, 9-oda
CHEBI:28801
CCCCCCCCC#CCCCCCCCC(=O)O

Specifications

189°C to 190°C (2mmHg)
1g
C18H32O2
1786634
RGTIBVZDHOMOKC-UHFFFAOYSA-N
octadec-9-ynoic acid
68167
280.45
9-Octadecynoic acid
43°C to 47°C
506-24-1
MFCD00014386
stearolic acid, 9-octadecynoic acid, 9-stearolic acid, delta 9-octadecynoic acid, stearolsaeure, octadec-9-insaeure, octadeca-9-ynoic acid, 9-odya, d09qia, 9-oda
CCCCCCCCC#CCCCCCCCC(=O)O
280.452
CHEBI:28801
98%