missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Nitroanthracene, tech. 85%

$85.91 - $85.91

Chemical Identifiers

CAS 602-60-8
Molecular Formula C14H9NO2
Molecular Weight (g/mol) 223.231
MDL Number MFCD00001248
InChI Key LSIKFJXEYJIZNB-UHFFFAOYSA-N
Synonym anthracene, 9-nitro, 5-nitroanthracene, unii-jz1tb3n57p, 9-nitro-anthracene, ccris 4679, jz1tb3n57p, 9-nitro anthracene, pubchem15119, acmc-20ab5c, 9-nitroanthracene
PubChem CID 11767
ChEBI CHEBI:82501
IUPAC Name 9-nitroanthracene
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2[N+](=O)[O-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0109306 View Documents Alfa Aesar™
L0109306
5g Each for $85.91
 
Specifications

Chemical Identifiers

602-60-8
223.231
LSIKFJXEYJIZNB-UHFFFAOYSA-N
11767
9-nitroanthracene
C14H9NO2
MFCD00001248
anthracene, 9-nitro, 5-nitroanthracene, unii-jz1tb3n57p, 9-nitro-anthracene, ccris 4679, jz1tb3n57p, 9-nitro anthracene, pubchem15119, acmc-20ab5c, 9-nitroanthracene
CHEBI:82501
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2[N+](=O)[O-]

Specifications

275°C (17mmHg)
5g
C14H9NO2
1877509
LSIKFJXEYJIZNB-UHFFFAOYSA-N
9-nitroanthracene
11767
223.23
Technical
138°C to 143°C
602-60-8
MFCD00001248
anthracene, 9-nitro, 5-nitroanthracene, unii-jz1tb3n57p, 9-nitro-anthracene, ccris 4679, jz1tb3n57p, 9-nitro anthracene, pubchem15119, acmc-20ab5c, 9-nitroanthracene
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2[N+](=O)[O-]
223.231
CHEBI:82501
85%
9-Nitroanthracene
Safety and Handling

Safety and Handling

RTECSNumber : CB0715000