Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Methylcarbazole-3-boronic acid pinacol ester, 95%

$63.43 - $63.43

Chemical Identifiers

Chemical Name or Material 9-Methylcarbazole-3-boronic acid pinacol ester
Molecular Formula C19H22BNO2
Formula Weight 307.2
MDL Number MFCD16294552
Synonym 9-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5195203

  SDS
alfa aesar™
H5195203
1g Each for $63.43

AAH5195206

  SDS
alfa aesar™
H5195206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 307.2
Quantity 1g
Assay 95%
MDL Number MFCD16294552
Molecular Formula C19H22BNO2
Synonym 9-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Chemical Name or Material 9-Methylcarbazole-3-boronic acid pinacol ester