missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Methylanthracene, TCI America™

Manufacturer:  TCI America M01535G

 View more versions of this product

Catalog No. M01535G



Specifications

Specifications

9-Methylanthracene
Undesignated
5g
C15H12
3077
CPGPAVAKSZHMBP-UHFFFAOYSA-N
9-methylanthracene
13068
Crystal-Powder at 20°C
370°C
79°C
779-02-2
MFCD00001261
anthracene, 9-methyl, unii-65nk4cin03, ccris 2740, 9-methyl anthracene, 9-methyl-anthracene, anthracen-9-ylmethyl, acmc-209pcm, 9-methylanthracene, wln: l c666j b1, 9-methylanthracene, analytical standard
CC1=C2C=CC=CC2=CC3=CC=CC=C13
192.261
192.26
≥98.0% (GC)

Chemical Identifiers

779-02-2
192.261
CPGPAVAKSZHMBP-UHFFFAOYSA-N
13068
CC1=C2C=CC=CC2=CC3=CC=CC=C13
C15H12
MFCD00001261
anthracene, 9-methyl, unii-65nk4cin03, ccris 2740, 9-methyl anthracene, 9-methyl-anthracene, anthracen-9-ylmethyl, acmc-209pcm, 9-methylanthracene, wln: l c666j b1, 9-methylanthracene, analytical standard
9-methylanthracene
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0640

RTECSNumber : CB0700000

TSCA : Yes

Documents