Learn More
9-Methylanthracene, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 126300050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
9-Methylanthracene | |
76.0°C to 79.0°C | |
1.0660g/mL | |
Authentic | |
Glass bottle | |
MFCD00001261 | |
05,II,586 | |
anthracene, 9-methyl, unii-65nk4cin03, ccris 2740, 9-methyl anthracene, 9-methyl-anthracene, anthracen-9-ylmethyl, acmc-209pcm, 9-methylanthracene, wln: l c666j b1, 9-methylanthracene, analytical standard | |
CC1=C2C=CC=CC2=CC2=CC=CC=C12 | |
192.26 | |
192.26 | |
Crystalline Powder or Crystals |
779-02-2 | |
Orange to Yellow | |
196.0°C to 197.0°C (12.0 mmHg) | |
98.5% min. (GC) | |
C15H12 | |
5 g | |
1.066 | |
CPGPAVAKSZHMBP-UHFFFAOYSA-N | |
9-methylanthracene | |
13068 | |
99% |
Chemical Identifiers
779-02-2 | |
192.26 | |
CPGPAVAKSZHMBP-UHFFFAOYSA-N | |
13068 | |
CC1=C2C=CC=CC2=CC2=CC=CC=C12 |
C15H12 | |
MFCD00001261 | |
anthracene, 9-methyl, unii-65nk4cin03, ccris 2740, 9-methyl anthracene, 9-methyl-anthracene, anthracen-9-ylmethyl, acmc-209pcm, 9-methylanthracene, wln: l c666j b1, 9-methylanthracene, analytical standard | |
9-methylanthracene |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
EINECSNumber : 212-299-7
RUO – Research Use Only