missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Methylanthracene, 99%

Manufacturer:  Alfa Aesar™ A1569914

 View more versions of this product

Catalog No. AAA1569914



Description

Description

Applications
9-Methylanthracene used as a Intermediates for pharmaceutical and chemical research.

Solubility
Soluble in water (0,0003 mg/L at 25°C) and toluene. Slightly soluble in benzene.

Notes
Mechanical exhaust required. When not in use, tightly seal the container and store in a dry, cool place.
Specifications

Specifications

9-Methylanthracene
77°C to 79°C
779-02-2
MFCD00001261
1907463
CPGPAVAKSZHMBP-UHFFFAOYSA-N
9-methylanthracene
13068
99%
196°C to 197°C (16mmHg)
25g
C15H12
UN3077
anthracene, 9-methyl, unii-65nk4cin03, ccris 2740, 9-methyl anthracene, 9-methyl-anthracene, anthracen-9-ylmethyl, acmc-209pcm, 9-methylanthracene, wln: l c666j b1, 9-methylanthracene, analytical standard
CC1=C2C=CC=CC2=CC3=CC=CC=C13
192.261
192.26

Chemical Identifiers

779-02-2
192.261
CPGPAVAKSZHMBP-UHFFFAOYSA-N
13068
CC1=C2C=CC=CC2=CC3=CC=CC=C13
C15H12
MFCD00001261
anthracene, 9-methyl, unii-65nk4cin03, ccris 2740, 9-methyl anthracene, 9-methyl-anthracene, anthracen-9-ylmethyl, acmc-209pcm, 9-methylanthracene, wln: l c666j b1, 9-methylanthracene, analytical standard
9-methylanthracene
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

GHS P Statement
P280
Wear protective gloves/protective clothing/eye protection/face protection.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : CB0700000

TSCA : TSCA

Documents