Please login to your online account to display your discounted pricing

9-Methyladenine, 98%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  325875000

 View more versions of this product

Catalog No. AC325875000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 149.17
Physical Form Crystals
Quantity 500mg
Color Beige
CAS Max % 100.0
Beilstein 26, V,16, 128
Synonym 6-Amino-9-methylpurine
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 9-Methyladenine
Name Note 98%
Molecular Formula C6H7N5
MDL Number MFCD00047232
CAS 700-00-5
Assay Percent Range 98%

Specifications

Chemical Name or Material 9-Methyladenine
Name Note 98%
CAS 700-00-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H7N5
Formula Weight 149.17
MDL Number MFCD00047232
Synonym 6-Amino-9-methylpurine
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents