Please login to your online account to display your discounted pricing

9-Methyladenine, 98%, ACROS Organics™

$833.07 - $833.07

Chemical Identifiers

Chemical Name or Material 9-Methyladenine
Name Note 98%
CAS 700-00-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H7N5
Formula Weight 149.17
MDL Number MFCD00047232
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC325875000

  SDS
thermo fisher scientific
325875000
Glass bottle 500mg Each for $833.07

AC325871000

  SDS
fisher scientific acros
325871000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystals
Formula Weight 149.17
Quantity 500mg
Color Beige
CAS Max % 100.0
Beilstein 26, V,16, 128
Chemical Name or Material 9-Methyladenine
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C6H7N5
MDL Number MFCD00047232
CAS 700-00-5
Synonym 6-Amino-9-methylpurine
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.