Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Isopropylcarbazole, 97%

$57.57 - $57.57

Chemical Identifiers

Chemical Name or Material 9-Isopropylcarbazole
CAS 1484-09-9
Molecular Formula C15H15N
Formula Weight 209.29
MDL Number MFCD00046896
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2764306

  SDS
alfa aesar™
H2764306
5g Each for $57.57
Description & Specifications

Specifications

Formula Weight 209.29
MDL Number MFCD00046896
TSCA TSCA
Assay 97%
Molecular Formula C15H15N
Melting Point 118° to 122°C
Chemical Name or Material 9-Isopropylcarbazole
CAS 1484-09-9
Quantity 5g