Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Hydroxy-9-fluorenecarboxylic acid, 97%

Chemical Identifiers

Chemical Name or Material 9-Hydroxy-9-fluorenecarboxylic acid
CAS 467-69-6
Molecular Formula C14H10O3
Formula Weight 226.23
MDL Number MFCD00001137
UN Number UN3077
Beilstein 1314711
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2240906

  SDS
alfa aesar™
B2240906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2240922

  SDS
alfa aesar™
B2240922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2240914

  SDS
alfa aesar™
B2240914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226.23
Chemical Name or Material 9-Hydroxy-9-fluorenecarboxylic acid
TSCA TSCA
Assay 97%
Molecular Formula C14H10O3
MDL Number MFCD00001137
CAS 467-69-6
UN Number UN3077
Beilstein 1314711
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number LL6480000
Melting Point ≈164°C (dec.)
Quantity 5g