Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Hydroxy-4-methoxyacridine, 97%

$89.80 - $89.80

Chemical Identifiers

Chemical Name or Material 9-Hydroxy-4-methoxyacridine
CAS 35308-00-0
Molecular Formula C14H11NO2
Formula Weight 225.25
MDL Number MFCD00005027
Beilstein 200297
Synonym 4-Methoxy-9-acridinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0135803

  SDS
alfa aesar™
L0135803
1g Each for $89.80
Description & Specifications

Specifications

Formula Weight 225.25
CAS 35308-00-0
Chemical Name or Material 9-Hydroxy-4-methoxyacridine
Synonym 4-Methoxy-9-acridinol
Beilstein 200297
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈286°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00005027
Health Hazard 1 Warning
Quantity 1g
Molecular Formula C14H11NO2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.