Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Hexadecyn-1-ol, 98%

$88.36 - $350.96

Chemical Identifiers

Chemical Name or Material 9-Hexadecyn-1-ol
CAS 88109-73-3
Molecular Formula C16H30O
Linear Formula CH3(CH2)5C≡C(CH2)8OH
Formula Weight 238.41
MDL Number MFCD00078285
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2172303

  SDS
alfa aesar™
B2172303
1g Each for $88.36

AAB2172306

  SDS
alfa aesar™
B2172306
5g Each for $350.96
Description & Specifications

Specifications

Formula Weight 238.41
Chemical Name or Material 9-Hexadecyn-1-ol
Assay 98%
Molecular Formula C16H30O
Linear Formula CH3(CH2)5C≡C(CH2)8OH
MDL Number MFCD00078285
CAS 88109-73-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
It is an important raw material and intermediate for pharmaceuticals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.