missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Heptadecanone, 98%
CAS: 540-08-9 | C17H34O | 254.458 g/mol
$129.59 - $434.26
Chemical Identifiers
| CAS | 540-08-9 |
|---|---|
| Molecular Formula | C17H34O |
| Molecular Weight (g/mol) | 254.458 |
| MDL Number | MFCD00009573 |
| InChI Key | WTJKUFMLQFLJOT-UHFFFAOYSA-N |
| Synonym | 9-heptadecanone, dioctyl ketone, pelargone, octyl ketone, di-n-octyl ketone, dioctylketone, 9-oxoheptadecane, bis-octyl ketone, di-n-octylketone, acmc-209lc2 |
| PubChem CID | 10887 |
| IUPAC Name | heptadecan-9-one |
| SMILES | CCCCCCCCC(=O)CCCCCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0163006
|
Thermo Scientific Chemicals
L0163006 |
5 g |
N/A
|
|
|||||
|
AAL0163014
|
Thermo Scientific Chemicals
L0163014 |
25 g |
N/A
|
|
|||||
Description
9-Heptadecanone is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications9-Heptadecanone is used as pharmaceutical intermediates.
Solubility
Soluble in toluene. Insoluble in water.
Notes
Avoid excessive heat and light. Always store away from incompatible compounds such as oxidizing agents.
Chemical Identifiers
| 540-08-9 | |
| 254.458 | |
| WTJKUFMLQFLJOT-UHFFFAOYSA-N | |
| 10887 | |
| CCCCCCCCC(=O)CCCCCCCC |
| C17H34O | |
| MFCD00009573 | |
| 9-heptadecanone, dioctyl ketone, pelargone, octyl ketone, di-n-octyl ketone, dioctylketone, 9-oxoheptadecane, bis-octyl ketone, di-n-octylketone, acmc-209lc2 | |
| heptadecan-9-one |
Specifications
| 540-08-9 | |
| 250°C to 253°C | |
| MFCD00009573 | |
| 1707117 | |
| Soluble in toluene. Insoluble in water. | |
| CCCCCCCCC(=O)CCCCCCCC | |
| 254.458 | |
| 254.46 | |
| 9-Heptadecanone |
| 51°C to 53°C | |
| C17H34O | |
| 5 g | |
| 9-heptadecanone, dioctyl ketone, pelargone, octyl ketone, di-n-octyl ketone, dioctylketone, 9-oxoheptadecane, bis-octyl ketone, di-n-octylketone, acmc-209lc2 | |
| WTJKUFMLQFLJOT-UHFFFAOYSA-N | |
| heptadecan-9-one | |
| 10887 | |
| 98% |
Safety and Handling
EINECSNumber : 208-734-5
RTECSNumber : MI3925000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only