Please login to your online account to display your discounted pricing

9-Fluorenylmethylecarbamate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 9-Fluorenylmethylecarbamate
CAS 84418-43-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (HPLC)
Assay Percent Range 99%
Molecular Formula C15H13NO2
Formula Weight 239.28
MDL Number MFCD00237376
Synonym FMOC-amide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC434170010

  SDS
thermo fisher scientific
AC434170010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC434170050

  SDS
thermo fisher scientific
AC434170050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 239.28
Physical Form Liquid
Assay ≥98.5% (HPLC)
Quantity 1g
Color Clear colorless to yellow
CAS Max % 100.0
Infrared Spectrum Authentic
Synonym FMOC-amide
CAS Min % 98.5
Assay Percent Range 99%
CAS 84418-43-9
Chemical Name or Material 9-Fluorenylmethylecarbamate
Molecular Formula C15H13NO2
MDL Number MFCD00237376
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.