Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Fluorenylmethyl pentafluorophenyl carbonate, 98%

$769.64 - $769.64

Chemical Identifiers

Chemical Name or Material 9-Fluorenylmethyl pentafluorophenyl carbonate
CAS 88744-04-1
Molecular Formula C21H11F5O3
Formula Weight 406.31
MDL Number MFCD00013261
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2229714

 
alfa aesar™
B2229714
25g Each for $769.64
Description & Specifications

Specifications

Formula Weight 406.31
Chemical Name or Material 9-Fluorenylmethyl pentafluorophenyl carbonate
Assay 98%
Molecular Formula C21H11F5O3
MDL Number MFCD00013261
CAS 88744-04-1
Recommended Storage 2° to 8°C
Melting Point 83° to 86°C
Quantity 25g
Safety and Handling

2° to 8°C