missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Fluorenylmethyl carbamate, 98+%

Manufacturer:  Alfa Aesar™ H5419906

 View more versions of this product

Catalog No. AAH5419906



Specifications

Specifications

9-Fluorenylmethyl carbamate
5g
C15H13NO2
4188192
ZZOKVYOCRSMTSS-UHFFFAOYSA-N
9H-fluoren-9-ylmethyl carbamate
736301
≥98%
201°C to 204°C
84418-43-9
MFCD00237376
9-fluorenemethane resin
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)N
239.274
239.27

Chemical Identifiers

84418-43-9
239.274
ZZOKVYOCRSMTSS-UHFFFAOYSA-N
736301
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)N
C15H13NO2
MFCD00237376
9-fluorenemethane resin
9H-fluoren-9-ylmethyl carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents