missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Fluorenylmethanol 98.0+%, TCI America™
Supplier: TCI America F020025G
Specifications
| 9-Fluorenylmethanol | |
| 105°C | |
| C14H12O | |
| 25 g | |
| XXSCONYSQQLHTH-UHFFFAOYSA-N | |
| (9H-fluoren-9-yl)methanol | |
| 90466 | |
| ≥98.0% (GC) |
| 24324-17-2 | |
| White-Yellow | |
| MFCD00001139 | |
| 9-fluorenemethanol, 9-fluorenylmethanol, 9h-fluorene-9-methanol, 9-fluorenyl methanol, 9h-fluoren-9-yl methanol, fluorene-9-methanol, 9-hydroxymethylfluorene, 9-fluorenmethanol, hofm, fluoren-9-yl methanol | |
| OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 196.25 | |
| 196.25 | |
| Crystalline Powder |
Chemical Identifiers
| 24324-17-2 | |
| 196.25 | |
| XXSCONYSQQLHTH-UHFFFAOYSA-N | |
| 90466 | |
| OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| C14H12O | |
| MFCD00001139 | |
| 9-fluorenemethanol, 9-fluorenylmethanol, 9h-fluorene-9-methanol, 9-fluorenyl methanol, 9h-fluoren-9-yl methanol, fluorene-9-methanol, 9-hydroxymethylfluorene, 9-fluorenmethanol, hofm, fluoren-9-yl methanol | |
| (9H-fluoren-9-yl)methanol |
Safety and Handling
TSCA : Yes