Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Fluorenylideneacetic acid, 99%

$74.45 - $910.08

Chemical Identifiers

Chemical Name or Material 9-Fluorenylideneacetic acid
CAS 4425-73-4
Molecular Formula C15H10O2
Formula Weight 222.24
MDL Number MFCD00040930
Synonym Dibenzofulvene-1-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1923206

  SDS
alfa aesar™
A1923206
5g Each for $74.45

AAA1923214

  SDS
alfa aesar™
A1923214
25g Each for $274.23

AAA1923222

  SDS
alfa aesar™
A1923222
100g Each for $910.08
Description & Specifications

Specifications

Formula Weight 222.24
Assay 99%
Molecular Formula C15H10O2
MDL Number MFCD00040930
CAS 4425-73-4
Synonym Dibenzofulvene-1-carboxylic acid
Melting Point 231° to 233°C
Quantity 5g
Chemical Name or Material 9-Fluorenylideneacetic acid