missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (9-Fluorenyl)triphenylphosphonium bromide, 98%

$42.72 - $125.23

Chemical Identifiers

CAS 7283-09-2
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00031650
InChI Key WQZGKKKJIJFFOK-UHFFFAOYNA-N
Synonym beta-d-allose, beta-d-allopyranose, 3 s-hydroxy-all-cis-8,11,14,17-eicosatetraenoyl-coa, 2r,3r,4r,5s,6r-6-hydroxymethyl oxane-2,3,4,5-tetrol, 1rpj, .beta.-d-allose, d +-allose, b-d-allopyranose, d-allose, beta-d-allose, rare aldohexose sugar
PubChem CID 448388
ChEBI CHEBI:40656
IUPAC Name (2R,3R,4R,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES C(C1C(C(C(C(O1)O)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0079403 View Documents Alfa Aesar™
L0079403
1g Each for $42.72
 
AAL0079406 View Documents Alfa Aesar™
L0079406
5g Each for $125.23
 
Specifications

Chemical Identifiers

7283-09-2
180.16
WQZGKKKJIJFFOK-UHFFFAOYNA-N
448388
(2R,3R,4R,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
C6H12O6
MFCD00031650
beta-d-allose, beta-d-allopyranose, 3 s-hydroxy-all-cis-8,11,14,17-eicosatetraenoyl-coa, 2r,3r,4r,5s,6r-6-hydroxymethyl oxane-2,3,4,5-tetrol, 1rpj, .beta.-d-allose, d +-allose, b-d-allopyranose, d-allose, beta-d-allose, rare aldohexose sugar
CHEBI:40656
C(C1C(C(C(C(O1)O)O)O)O)O

Specifications

>330°C
7253-07-8
MFCD00031650
9-fluorenyl triphenylphosphonium bromide, 9h-fluoren-9-yltriphenylphosphanium bromide, 9-fluorenyltriphenylphosphium bromide, 9h-fluoren-9-yl triphenylphosphonium bromide
C1=CC=C(C=C1)[P+](C2C3=CC=CC=C3C4=CC=CC=C24)(C5=CC=CC=C5)C6=CC=CC=C6.[Br-]
507.41
507.42
(9-Fluorenyl)triphenylphosphonium bromide
1g
C31H24BrP
3897970
BHNPKGAMAZKCCJ-UHFFFAOYSA-M
9H-fluoren-9-yl(triphenyl)phosphanium;bromide
18702719
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning