Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Fluorenone hydrazone, 98+%

$41.55 - $103.76

Chemical Identifiers

Chemical Name or Material 9-Fluorenone hydrazone
CAS 13629-22-6
Molecular Formula C13H10N2
Formula Weight 194.24
MDL Number MFCD00016357
Beilstein 1910575
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2170606

 
alfa aesar™
B2170606
5g Each for $41.55

AAB2170614

 
alfa aesar™
B2170614
25g Each for $103.76
Description & Specifications

Specifications

Formula Weight 194.24
Chemical Name or Material 9-Fluorenone hydrazone
Assay ≥98%
Molecular Formula C13H10N2
MDL Number MFCD00016357
CAS 13629-22-6
Beilstein 1910575
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 150° to 154°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.