missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
9-Fluorenone hydrazone, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC119230050
Specifications
9-Fluorenone hydrazone | |
148.0°C to 150.0°C | |
13629-22-6 | |
C13H10N2 | |
9-fluorenone hydrazone, 9h-fluoren-9-one, hydrazone, 9h-fluoren-9-ylidene hydrazine, 9h-fluoren-9-ylidenehydrazine, fluoren-9-one hydrazone, fluoren-9-one, hydrazone, fluoren-9-ylidene-hydrazine, fluoren-9-ylidenediazane, fluorenone hydrazone, fluorenonhydrazon | |
YCNUILAKOMIBAL-UHFFFAOYSA-N | |
fluoren-9-ylidenehydrazine | |
83627 | |
97% |
Glass bottle | |
5g | |
96% min. (HPLC) | |
07,I,251 | |
Solubility in water: insoluble | |
C1=CC=C2C(=C1)C3=CC=CC=C3C2=NN | |
194.24 | |
194.24 |
Chemical Identifiers
13629-22-6 | |
194.24 | |
9-fluorenone hydrazone, 9h-fluoren-9-one, hydrazone, 9h-fluoren-9-ylidene hydrazine, 9h-fluoren-9-ylidenehydrazine, fluoren-9-one hydrazone, fluoren-9-one, hydrazone, fluoren-9-ylidene-hydrazine, fluoren-9-ylidenediazane, fluorenone hydrazone, fluorenonhydrazon | |
fluoren-9-ylidenehydrazine |
C13H10N2 | |
YCNUILAKOMIBAL-UHFFFAOYSA-N | |
83627 | |
C1=CC=C2C(=C1)C3=CC=CC=C3C2=NN |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 237-116-8