missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Fluorenone hydrazone, 97%, ACROS Organics™

Chemical Identifiers

CAS 13629-22-6
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.24
InChI Key YCNUILAKOMIBAL-UHFFFAOYSA-N
Synonym 9-fluorenone hydrazone, 9h-fluoren-9-one, hydrazone, 9h-fluoren-9-ylidene hydrazine, 9h-fluoren-9-ylidenehydrazine, fluoren-9-one hydrazone, fluoren-9-one, hydrazone, fluoren-9-ylidene-hydrazine, fluoren-9-ylidenediazane, fluorenone hydrazone, fluorenonhydrazon
PubChem CID 83627
IUPAC Name fluoren-9-ylidenehydrazine
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2=NN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC119230050
SDS View Documents
Thermo Fisher Scientific
AC119230050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC119230250
View Documents
Thermo Fisher Scientific
AC119230250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

13629-22-6
194.24
9-fluorenone hydrazone, 9h-fluoren-9-one, hydrazone, 9h-fluoren-9-ylidene hydrazine, 9h-fluoren-9-ylidenehydrazine, fluoren-9-one hydrazone, fluoren-9-one, hydrazone, fluoren-9-ylidene-hydrazine, fluoren-9-ylidenediazane, fluorenone hydrazone, fluorenonhydrazon
fluoren-9-ylidenehydrazine
C13H10N2
YCNUILAKOMIBAL-UHFFFAOYSA-N
83627
C1=CC=C2C(=C1)C3=CC=CC=C3C2=NN

Specifications

Glass bottle
5g
96% min. (HPLC)
07,I,251
Solubility in water: insoluble
C1=CC=C2C(=C1)C3=CC=CC=C3C2=NN
194.24
194.24
9-Fluorenone hydrazone
148.0°C to 150.0°C
13629-22-6
C13H10N2
9-fluorenone hydrazone, 9h-fluoren-9-one, hydrazone, 9h-fluoren-9-ylidene hydrazine, 9h-fluoren-9-ylidenehydrazine, fluoren-9-one hydrazone, fluoren-9-one, hydrazone, fluoren-9-ylidene-hydrazine, fluoren-9-ylidenediazane, fluorenone hydrazone, fluorenonhydrazon
YCNUILAKOMIBAL-UHFFFAOYSA-N
fluoren-9-ylidenehydrazine
83627
97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 237-116-8

SDS