Please login to your online account to display your discounted pricing

9-Fluorenone, 99+%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: fisher scientific acros  119200050

 View more versions of this product

Catalog No. AC119200050

  • / Each
 

Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass Bottle
Melting Point 81° to 85°C
Color Yellow
Quantity 5g
Chemical Name or Material 9-Fluorenone
CAS 486-25-9
Physical Form Yellow crystalline powder or flakes
Assay 99.0% min.
Assay Percent Range 99+%
Molecular Formula C13H8O
Formula Weight 180.21g/mol

Specifications

Chemical Name or Material 9-Fluorenone
Assay 99.0% min.
Assay Percent Range 99+%
Molecular Formula C13H8O
Formula Weight 180.21g/mol