Please login to your online account to display your discounted pricing

9-Fluorenone-2-carboxylic Acid 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 9-Fluorenone-2-carboxylic acid
Name Note 97%
CAS 784-50-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C14H8O3
Formula Weight 224.22
MDL Number MFCD00001156
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC119210010

  SDS
fisher scientific acros
119210010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 224.22
Physical Form Fine Powder
Quantity 1g
Color Yellow
EINECS Number 212-317-3
Beilstein 10, 774
CAS Min % 96.0
CAS Max % 100.0
Synonym 9-Oxo-2-fluorenecarboxylic acid
Packaging Glass bottle
Infrared Spectrum Authentic
Name Note 97%
CAS 784-50-9
Chemical Name or Material 9-Fluorenone-2-carboxylic acid
MDL Number MFCD00001156
Assay Percent Range 97%
Molecular Formula C14H8O3