Please login to your online account to display your discounted pricing

9-Fluorenone-1-carboxylic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 9-Fluorenone-1-carboxylic acid
CAS 1573-92-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H8O3
Formula Weight 224.22
MDL Number MFCD00011537
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC373350050

  SDS
thermo fisher scientific
AC373350050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 224.22
Quantity 5g
Color Orange
Melting Point 194° to 198°C
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00011537
CAS 1573-92-8
Molecular Formula C14H8O3
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
CAS Min % 97.5
Infrared Spectrum Authentic
Chemical Name or Material 9-Fluorenone-1-carboxylic acid
Assay Percent Range 98%