missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Fluorenone-1-carboxylic acid, 98%

$87.43 - $246.28

Chemical Identifiers

CAS 1573-92-8
Molecular Formula C14H8O3
Formula Weight 224.22
MDL Number MFCD00011537
Chemical Name or Material 9-Fluorenone-1-carboxylic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2028303 SDS Alfa Aesar™
B2028303
1g Each for $87.43
AAB2028306 SDS Alfa Aesar™
B2028306
5g Each for $246.28
Specifications Description & Specifications

Chemical Identifiers

1573-92-8
224.22
9-Fluorenone-1-carboxylic acid
C14H8O3
MFCD00011537

Specifications

196° to 198°C
1573-92-8
224.22
9-Fluorenone-1-carboxylic acid
1g
C14H8O3
MFCD00011537
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning