missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Fluorenol, TCI America™

Manufacturer:  TCI America F0229250G

 View more versions of this product

Catalog No. F0229250G



Specifications

Specifications

9-Fluorenol
155°C
1689-64-1
MFCD00001135
9-hydroxyfluorene, 9-fluorenol, fluoren-9-ol, fluorenol, 9-hydroxy fluorene, diphenylene carbinol, unii-bv0q72r613, hydroxyfluoren, 9-hydroxy-fluoren, 9h-fluorene-9-ol
C1=CC=C2C(=C1)C(C3=CC=CC=C32)O
182.222
CHEBI:16904
Crystal-Powder at 20°C
White-Yellow
250g
C13H10O
3077
AFMVESZOYKHDBJ-UHFFFAOYSA-N
9H-fluoren-9-ol
74318
182.22
≥99.0% (GC)

Chemical Identifiers

1689-64-1
182.222
AFMVESZOYKHDBJ-UHFFFAOYSA-N
74318
9H-fluoren-9-ol
C13H10O
MFCD00001135
9-hydroxyfluorene, 9-fluorenol, fluoren-9-ol, fluorenol, 9-hydroxy fluorene, diphenylene carbinol, unii-bv0q72r613, hydroxyfluoren, 9-hydroxy-fluoren, 9h-fluorene-9-ol
CHEBI:16904
C1=CC=C2C(=C1)C(C3=CC=CC=C32)O
Safety and Handling

Safety and Handling

TSCA : Yes

Documents