missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
9-Fluoreneacetic Acid 98.0+%, TCI America™
Supplier: TCI America F03301G
Specifications
9-Fluoreneacetic Acid | |
Yellow | |
MFCD00013262 | |
9-fluoreneacetic acid, fluorene-9-acetic acid, 9h-fluorene-9-acetic acid, 2-9h-fluoren-9-yl acetic acid, 9h-fluorene-9-aceticacid, 9h-fluoren-9-ylacetic acid, chembl44381, 2-fluoren-9-ylacetic acid, 9-fluoreneacetate, 9-fluorene acetic acid | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)CC(=O)O | |
224.259 | |
224.26 | |
Crystalline Powder |
6284-80-6 | |
C15H12O2 | |
1 g | |
WFSMJMTYIMFHPV-UHFFFAOYSA-N | |
2-(9H-fluoren-9-yl)acetic acid | |
94852 | |
≥98.0% (T) |
Chemical Identifiers
6284-80-6 | |
224.259 | |
WFSMJMTYIMFHPV-UHFFFAOYSA-N | |
94852 | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)CC(=O)O |
C15H12O2 | |
MFCD00013262 | |
9-fluoreneacetic acid, fluorene-9-acetic acid, 9h-fluorene-9-acetic acid, 2-9h-fluoren-9-yl acetic acid, 9h-fluorene-9-aceticacid, 9h-fluoren-9-ylacetic acid, chembl44381, 2-fluoren-9-ylacetic acid, 9-fluoreneacetate, 9-fluorene acetic acid | |
2-(9H-fluoren-9-yl)acetic acid |
Safety and Handling
TSCA : No