missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Ethylcarbazole-3-boronic acid, 98%

$29.45 - $345.43

Chemical Identifiers

CAS 669072-93-9
Molecular Formula C14H14BNO2
Molecular Weight (g/mol) 239.081
MDL Number MFCD09743121
InChI Key JUBKLYCRFZTXBA-UHFFFAOYSA-N
Synonym 9-ethyl-9h-carbazol-3-yl boronic acid, 9-ethyl-3-carbazole boronic acid, boronic acid, 9-ethyl-9h-carbazol-3-yl, ksc924s3d, 9-ethylcarbazole-3-boronic acid, 9-ethyl-9h-carbazol-3-yl boronicacid
PubChem CID 45382262
IUPAC Name (9-ethylcarbazol-3-yl)boronic acid
SMILES B(C1=CC2=C(C=C1)N(C3=CC=CC=C32)CC)(O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64959MD View Documents Alfa Aesar™
H64959MD
250mg Each for $29.45
 
AAH6495903 View Documents Alfa Aesar™
H6495903
1g Each for $86.99
 
AAH6495906 View Documents Alfa Aesar™
H6495906
5g Each for $345.43
 
Specifications

Chemical Identifiers

669072-93-9
239.081
JUBKLYCRFZTXBA-UHFFFAOYSA-N
45382262
B(C1=CC2=C(C=C1)N(C3=CC=CC=C32)CC)(O)O
C14H14BNO2
MFCD09743121
9-ethyl-9h-carbazol-3-yl boronic acid, 9-ethyl-3-carbazole boronic acid, boronic acid, 9-ethyl-9h-carbazol-3-yl, ksc924s3d, 9-ethylcarbazole-3-boronic acid, 9-ethyl-9h-carbazol-3-yl boronicacid
(9-ethylcarbazol-3-yl)boronic acid

Specifications

250mg
C14H14BNO2
23374049
JUBKLYCRFZTXBA-UHFFFAOYSA-N
(9-ethylcarbazol-3-yl)boronic acid
45382262
Solid
9-Ethylcarbazole-3-boronic acid
669072-93-9
MFCD09743121
9-ethyl-9h-carbazol-3-yl boronic acid, 9-ethyl-3-carbazole boronic acid, boronic acid, 9-ethyl-9h-carbazol-3-yl, ksc924s3d, 9-ethylcarbazole-3-boronic acid, 9-ethyl-9h-carbazol-3-yl boronicacid
B(C1=CC2=C(C=C1)N(C3=CC=CC=C32)CC)(O)O
239.081
239.08
98%