missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-cis-Retinoic acid, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals J62219LB0

Catalog No. AAJ62219LB0



Description

Description

9-cis-Retinoic acid is a vitamin A analog that inhibits cell proliferation and induces cell differentiation. 9-cis-Retinoic acid is an inhibitor of Cox-2 and an activator of RAR α, RAR β, RAR γ and RXR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-cis-Retinoic acid is a vitamin A analog that inhibits cell proliferation and induces cell differentiation. 9-cis-Retinoic acid is an inhibitor of Cox-2 and an activator of RAR α, RAR β, RAR γ and RXR.

Solu
Specifications

Specifications

9-cis-Retinoic acid
178°C to 180°C
MFCD00270072
Light and moisture sensitive
alitretinoin, 9-cis-retinoic acid, panretin, 9-cis-tretinoin, panrexin, panretyn, panretin gel, toctino, 9-cis retinoic acid, 9 z-retinoic acid
SHGAZHPCJJPHSC-YCNIQYBTSA-N
(2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
449171
300.44
5300-03-8
C20H28O2
5 mg
14,249
Soluble in chloroform (50mg/ml),DMSO (∽25mg/ml),ethanol (25mg/ml),and acetone (25mg/ml). Insoluble in water.
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C
300.44
CHEBI:50648
Powder

Chemical Identifiers

5300-03-8
300.44
SHGAZHPCJJPHSC-YCNIQYBTSA-N
449171
(2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
C20H28O2
MFCD00270072
alitretinoin, 9-cis-retinoic acid, panretin, 9-cis-tretinoin, panrexin, panretyn, panretin gel, toctino, 9-cis retinoic acid, 9 z-retinoic acid
CHEBI:50648
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C
Safety and Handling

Safety and Handling

GHS H Statement
H360-H315-H319-H335
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P271-P281-P304+P340-P308+P313-P501c

H335-H360D

RTECSNumber : VH6450000

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only