missing translation for 'onlineSavingsMsg'
Learn More

9-(Chloromethyl)anthracene, 98+%

CAS: 24463-19-2 | C15H11Cl | 226.703 g/mol

Supplier:  Thermo Scientific Chemicals A1357914

 View more versions of this product

Catalog No. AAA1357914


Only null left
Explore available promotions

Description

Description

9-(Chloromethyl)anthracene is used as a blocking group reagent used for carboxylic acids, phenols, mercaptans and thiophenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-(Chloromethyl)anthracene is used as a blocking group reagent used for carboxylic acids, phenols, mercaptans and thiophenols.

Solubility
Soluble in chloroform. Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

9-(Chloromethyl)anthracene
138°C to 142°C
MFCD00001263
UN3261
9-chloromethyl anthracene, anthracene, 9-chloromethyl, 9-anthracenylmethyl chloride, ccris 7993, 9-chloromethyanthracene, pubchem14854, 9-chlormethyl-anthracen, acmc-209tez
PCVRSXXPGXRVEZ-UHFFFAOYSA-N
9-(chloromethyl)anthracene
32385
≥98%
24463-19-2
C15H11Cl
25 g
1873394
Soluble in chloroform. Insoluble in water.
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CCl
226.703
226.71

Chemical Identifiers

24463-19-2
226.703
PCVRSXXPGXRVEZ-UHFFFAOYSA-N
32385
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CCl
C15H11Cl
MFCD00001263
9-chloromethyl anthracene, anthracene, 9-chloromethyl, 9-anthracenylmethyl chloride, ccris 7993, 9-chloromethyanthracene, pubchem14854, 9-chlormethyl-anthracen, acmc-209tez
9-(chloromethyl)anthracene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 246-274-7

RTECSNumber : CA9600000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only