Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Chlorofluorene, 98%

$106.99 - $462.58

Chemical Identifiers

Chemical Name or Material 9-Chlorofluorene
CAS 6630-65-5
Molecular Formula C13H9Cl
Formula Weight 200.67
MDL Number MFCD00043468
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5035203

  SDS
alfa aesar™
H5035203
1g Each for $106.99

AAH5035206

  SDS
alfa aesar™
H5035206
5g Each for $462.58
Description & Specifications

Specifications

Formula Weight 200.67
CAS 6630-65-5
Chemical Name or Material 9-Chlorofluorene
Assay 98%
Quantity 1g
RTECS Number LL6930000
MDL Number MFCD00043468
Melting Point 86° to 88°C
Molecular Formula C13H9Cl