missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Bromotriptycene 98.0+%, TCI America™

$197.13 - $695.34

Chemical Identifiers

CAS 15364-55-3
Molecular Formula C20H13Br
Molecular Weight (g/mol) 333.23
MDL Number MFCD01099022
InChI Key HSROKCVTEYMWHO-UHFFFAOYSA-N
Synonym 9-Bromo-9,10-dihydro-9,10-[1,2]benzenoanthracene
PubChem CID 139927
IUPAC Name 1-bromopentacyclo[6.6.6.0²,⁷.0⁹,¹⁴.0¹⁵,²⁰]icosa-2,4,6,9,11,13,15,17,19-nonaene
SMILES BrC12C3=CC=CC=C3C(C3=CC=CC=C13)C1=CC=CC=C21
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B39551G
View Documents
Tci America
B39551G
1g
Each for $197.13
 
B39555G
View Documents
Tci America
B39555G
5g
Each for $695.34
 
Description

Description

Specifications

Chemical Identifiers

15364-55-3
333.23
HSROKCVTEYMWHO-UHFFFAOYSA-N
139927
BrC12C3=CC=CC=C3C(C3=CC=CC=C13)C1=CC=CC=C21
C20H13Br
MFCD01099022
9-Bromo-9,10-dihydro-9,10-[1,2]benzenoanthracene
1-bromopentacyclo[6.6.6.0²,⁷.0⁹,¹⁴.0¹⁵,²⁰]icosa-2,4,6,9,11,13,15,17,19-nonaene

Specifications

Yellow
1g
C20H13Br
9-Bromo-9,10-dihydro-9,10-[1,2]benzenoanthracene
BrC12C3=CC=CC=C3C(C3=CC=CC=C13)C1=CC=CC=C21
333.23
333.23
≥98.0% (GC)
254°C
15364-55-3
MFCD01099022
HSROKCVTEYMWHO-UHFFFAOYSA-N
1-bromopentacyclo[6.6.6.0²,⁷.0⁹,¹⁴.0¹⁵,²⁰]icosa-2,4,6,9,11,13,15,17,19-nonaene
139927
Crystal-Powder at 20°C
9-Bromotriptycene
Safety and Handling

Safety and Handling

TSCA : No

SDS
Product Certifications