missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Bromononanoic Acid, TCI America™

$62.64 - $220.00

Chemical Identifiers

CAS 41059-02-3
Molecular Formula C9H17BrO2
Molecular Weight (g/mol) 237.137
MDL Number MFCD00046564
InChI Key XEGRKZRPTBNSMN-UHFFFAOYSA-N
PubChem CID 548221
IUPAC Name 9-bromononanoic acid
SMILES C(CCCCBr)CCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B23231G View Documents TCI America
B23231G
1g Each for $62.64
 
B23235G View Documents TCI America
B23235G
5g Each for $220.00
 
Specifications

Chemical Identifiers

41059-02-3
237.137
XEGRKZRPTBNSMN-UHFFFAOYSA-N
9-bromononanoic acid
C9H17BrO2
MFCD00046564
548221
C(CCCCBr)CCCC(=O)O

Specifications

174°C
37°C
41059-02-3
MFCD00046564
XEGRKZRPTBNSMN-UHFFFAOYSA-N
9-bromononanoic acid
548221
Crystal-Powder at 20°C
9-Bromononanoic Acid
Undesignated
1g
C9H17BrO2
3261
C(CCCCBr)CCCC(=O)O
237.137
237.14
≥97.0% (T)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator