missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Bromoanthracene 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 106640250

 View more versions of this product

Catalog No. AC106640250


 

Specifications Description & Specifications

Specifications

9-Bromoanthracene
1564-64-3
100.0
Glass bottle
≥95%
C14H9Br
MFCD00001243
Yellow
96%
95.0
Authentic
Powder
96%
257.13
05, 665
25g

Chemical Identifiers

1564-64-3
100.0
C14H9Br
MFCD00001243
≥95%
95.0
96%
257.13
9-Bromoanthracene, 96%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 216-359-3