missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Bromoanthracene, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0673303

 View more versions of this product

Catalog No. AAL0673303



Description

Description

9-Bromoanthracene acts as an intermediate in the preparation of 9-substituted derivative of the polycyclic aromatic hydrocarbon (PAH) anthracene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Bromoanthracene acts as an intermediate in the preparation of 9-substituted derivative of the polycyclic aromatic hydrocarbon (PAH) anthracene.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

9-Bromoanthracene
97°C to 103°C
MFCD00001243
1869747
Insoluble in water.
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2Br
257.13
257.14
1564-64-3
C14H9Br
1 g
anthracene, 9-bromo, 9-bromo anthracene, 9-bromo-anthracene, 9-anthracenyl bromide, 9-bromanthracen, pubchem9183, 9-bromoanthrene, acmc-209ddx, 9-bromoanthracene, dsstox_cid_29080
ZIRVQSRSPDUEOJ-UHFFFAOYSA-N
9-bromoanthracene
74062
96%

Chemical Identifiers

1564-64-3
257.13
ZIRVQSRSPDUEOJ-UHFFFAOYSA-N
74062
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2Br
C14H9Br
MFCD00001243
anthracene, 9-bromo, 9-bromo anthracene, 9-bromo-anthracene, 9-anthracenyl bromide, 9-bromanthracen, pubchem9183, 9-bromoanthrene, acmc-209ddx, 9-bromoanthracene, dsstox_cid_29080
9-bromoanthracene
Safety and Handling

Safety and Handling

P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H335-H340

EINECSNumber : 216-359-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only