missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Bromo-9-phenylfluorene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC428610050

 View more versions of this product

Catalog No. AC428610050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

9-Bromo-9-phenylfluorene
99°C to 101°C
55135-66-5
100.0
C19H13Br
9-brom-9-phenyl-9h-fluoren, 9-bromanyl-9-phenyl-fluorene, 9-bromo-9-phenyl-9h-fluorene, 9-bromo-9-phenyl-fluorene, 9-bromo-9-phenylfluorene, 9h-fluorene, 9-bromo-9-phenyl, acmc-209ll7, hyqxncdbsalqlb-uhfffaoysa, zlchem 855
HYQXNCDBSALQLB-UHFFFAOYSA-N
9-bromo-9-phenylfluorene
231624
97%
Glass bottle
5g
96.0
96% min. (HPLC)
MFCD00075522
Solubility in water: insoluble
C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)Br
321.21
321.21

Chemical Identifiers

55135-66-5
321.21
HYQXNCDBSALQLB-UHFFFAOYSA-N
231624
C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)Br
C19H13Br
MFCD00075522
9-brom-9-phenyl-9h-fluoren, 9-bromanyl-9-phenyl-fluorene, 9-bromo-9-phenyl-9h-fluorene, 9-bromo-9-phenyl-fluorene, 9-bromo-9-phenylfluorene, 9h-fluorene, 9-bromo-9-phenyl, acmc-209ll7, hyqxncdbsalqlb-uhfffaoysa, zlchem 855
9-bromo-9-phenylfluorene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents