missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Bromo-9,10-dihydro-9,10-ethenoanthracene, TCI America™

$345.28 - $1,208.50

Chemical Identifiers

CAS 126690-96-8
Molecular Formula C16H11Br
Molecular Weight (g/mol) 283.168
InChI Key JNPBTNVWRDISGS-UHFFFAOYSA-N
Synonym 9-Bromodibenzobarrelene
PubChem CID 91972153
SMILES C1=CC=C2C(=C1)C3C=CC2(C4=CC=CC=C34)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B42391G View Documents TCI America
B42391G
1g Each for $345.28
 
B42395G View Documents TCI America
B42395G
5g Each for $1,208.50
 
Specifications

Chemical Identifiers

126690-96-8
283.168
9-Bromodibenzobarrelene
C1=CC=C2C(=C1)C3C=CC2(C4=CC=CC=C34)Br
C16H11Br
JNPBTNVWRDISGS-UHFFFAOYSA-N
91972153

Specifications

White-Yellow
1g
C16H11Br
JNPBTNVWRDISGS-UHFFFAOYSA-N
283.168
283.17
≥98.0% (GC)
140°C
126690-96-8
9-Bromodibenzobarrelene
C1=CC=C2C(=C1)C3C=CC2(C4=CC=CC=C34)Br
91972153
Crystal-Powder at 20°C
9-Bromo-9,10-dihydro-9,10-ethenoanthracene
Safety and Handling

Safety and Handling

TSCA : No